Product Details
- SNP ID
-
rs75086416
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:7770123 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGAACGAGAAAAAGAAGAAAAAAA[C/T]GAAAAGGAGCTGCAAGATGAACAGG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NANOGNB
PubMed Links
Gene Details
- Gene
- NANOGNB
- Gene Name
- NANOG neighbor homeobox
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001145465.1 |
313 |
Silent Mutation |
AAC,AAT |
N81N |
NP_001138937.1 |
View Full Product Details