Product Details

SNP ID
rs76277635
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122208019 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTAAATCATTTTTGACGACGTAA[A/G]TAAGACTGAAAACAGGTTAAACAGT
Phenotype
MIM: 605864 MIM: 605219
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
B3GNT4 PubMed Links

Gene Details

Gene
B3GNT4
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011538732.1 1823 UTR 3 XP_011537034.1
Gene
DIABLO
Gene Name
diablo IAP-binding mitochondrial protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278302.1 1823 UTR 3 NP_001265231.1
NM_001278303.1 1823 UTR 3 NP_001265232.1
NM_001278304.1 1823 UTR 3 NP_001265233.1
NM_001278342.1 1823 UTR 3 NP_001265271.1
NM_019887.5 1823 UTR 3 NP_063940.1
NM_138930.3 1823 UTR 3 NP_620308.1
Gene
LRRC43
Gene Name
leucine rich repeat containing 43
There are no transcripts associated with this gene.

View Full Product Details