Product Details

SNP ID
rs74639204
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:1567155 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCACAGCCACTATCAAGAAAAAGA[C/T]ATAAGACATACTAAAGGGCAAATAG
Phenotype
MIM: 609081
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXL14 PubMed Links

Gene Details

Gene
FBXL14
Gene Name
F-box and leucine rich repeat protein 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152441.2 Intron NP_689654.1
XM_017018875.1 Intron XP_016874364.1
XM_017018876.1 Intron XP_016874365.1

View Full Product Details