Product Details

SNP ID
rs76684608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:5562748 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTAGCTCCAAACGTTTTTGTGA[C/T]GTGAAGTGGAGACAAGCCCAGGTTT
Phenotype
MIM: 610109
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANO2 PubMed Links

Gene Details

Gene
ANO2
Gene Name
anoctamin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278596.1 3880 UTR 3 NP_001265525.1
NM_001278597.1 3880 UTR 3 NP_001265526.1
XM_006718988.3 3880 UTR 3 XP_006719051.1
XM_011520975.2 3880 UTR 3 XP_011519277.1
XM_011520978.2 3880 UTR 3 XP_011519280.1
XM_017019672.1 3880 Intron XP_016875161.1

View Full Product Details