Product Details

SNP ID
rs77651612
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6667185 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAATCTCCTTCTACCAGCAGTCAA[C/T]AGGAAGCAAAGTGAGACGATGTAGG
Phenotype
MIM: 608524 MIM: 609951
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ING4 PubMed Links

Gene Details

Gene
ING4
Gene Name
inhibitor of growth family member 4
There are no transcripts associated with this gene.

Gene
ZNF384
Gene Name
zinc finger protein 384
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039920.2 2603 UTR 3 NP_001035009.1
NM_001135734.2 2603 UTR 3 NP_001129206.1
NM_133476.4 2603 UTR 3 NP_597733.2
XM_006718969.3 2603 UTR 3 XP_006719032.1
XM_011520917.1 2603 UTR 3 XP_011519219.1
XM_011520918.1 2603 UTR 3 XP_011519220.1
XM_011520919.1 2603 UTR 3 XP_011519221.1
XM_011520920.1 2603 UTR 3 XP_011519222.1
XM_011520921.2 2603 UTR 3 XP_011519223.1
XM_011520922.1 2603 UTR 3 XP_011519224.1
XM_011520923.1 2603 UTR 3 XP_011519225.1
XM_017018941.1 2603 UTR 3 XP_016874430.1
XM_017018942.1 2603 UTR 3 XP_016874431.1
XM_017018943.1 2603 UTR 3 XP_016874432.1
XM_017018944.1 2603 UTR 3 XP_016874433.1
XM_017018945.1 2603 UTR 3 XP_016874434.1
XM_017018946.1 2603 UTR 3 XP_016874435.1
XM_017018947.1 2603 UTR 3 XP_016874436.1
XM_017018948.1 2603 UTR 3 XP_016874437.1
XM_017018949.1 2603 UTR 3 XP_016874438.1
XM_017018950.1 2603 UTR 3 XP_016874439.1
XM_017018951.1 2603 UTR 3 XP_016874440.1
XM_017018952.1 2603 UTR 3 XP_016874441.1
XM_017018953.1 2603 UTR 3 XP_016874442.1
XM_017018954.1 2603 UTR 3 XP_016874443.1

View Full Product Details