Product Details

SNP ID
rs74516777
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:119674023 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGAGAGCATCAGTACAAGTTCT[C/T]TGTGGATGGTCAGTGGACGCACGAC
Phenotype
MIM: 602740
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105370027 PubMed Links

Gene Details

Gene
LOC105370027
Gene Name
uncharacterized LOC105370027
There are no transcripts associated with this gene.

Gene
PRKAB1
Gene Name
protein kinase AMP-activated non-catalytic subunit beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006253.4 574 Missense Mutation TCT,TTT S128F NP_006244.2
XM_005253909.1 574 Missense Mutation TCT,TTT S128F XP_005253966.1

View Full Product Details