Product Details

SNP ID
rs77408535
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:121809208 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTTAAATCCTAAAGCACTGGGT[A/G]ACTAGGAGGACAAAGCCCTTGGCCA
Phenotype
MIM: 611055
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC01089 PubMed Links

Gene Details

Gene
LINC01089
Gene Name
long intergenic non-protein coding RNA 1089
There are no transcripts associated with this gene.

Gene
SETD1B
Gene Name
SET domain containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015048.1 Intron NP_055863.1
XM_005253858.4 Intron XP_005253915.1
XM_006719296.3 Intron XP_006719359.1

View Full Product Details