Product Details

SNP ID
rs74630711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:109574285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACGAGGCGTACTGGGTGTTTTCT[A/G]CAGCCTTGTTTATAATTGTGACACA
Phenotype
MIM: 607568 MIM: 251170
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MMAB PubMed Links

Gene Details

Gene
MMAB
Gene Name
methylmalonic aciduria (cobalamin deficiency) cblB type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052845.3 255 Intron NP_443077.1
XM_011538267.2 255 Intron XP_011536569.1
XM_011538268.2 255 Intron XP_011536570.1
XM_011538269.2 255 Intron XP_011536571.1
Gene
MVK
Gene Name
mevalonate kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000431.3 255 Intron NP_000422.1
NM_001114185.2 255 Intron NP_001107657.1
NM_001301182.1 255 Intron NP_001288111.1
XM_011538372.2 255 Intron XP_011536674.2
XM_017019313.1 255 Intron XP_016874802.1
XM_017019314.1 255 UTR 5 XP_016874803.1

View Full Product Details