Product Details

SNP ID
rs79157363
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:109783135 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTGAGCACCGGCAAATCCCAGAC[A/G]CTACAGAACACACAGAAGGCATGGC
Phenotype
MIM: 605427
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM222A-AS1 PubMed Links

Gene Details

Gene
FAM222A-AS1
Gene Name
FAM222A antisense RNA 1
There are no transcripts associated with this gene.

Gene
TRPV4
Gene Name
transient receptor potential cation channel subfamily V member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177428.1 3000 UTR 3 NP_001170899.1
NM_001177431.1 3000 UTR 3 NP_001170902.1
NM_001177433.1 3000 UTR 3 NP_001170904.1
NM_021625.4 3000 UTR 3 NP_067638.3
NM_147204.2 3000 UTR 3 NP_671737.1
XM_005253918.1 3000 UTR 3 XP_005253975.1
XM_011538630.2 3000 UTR 3 XP_011536932.2
XM_011538631.2 3000 UTR 3 XP_011536933.2
XM_011538632.2 3000 UTR 3 XP_011536934.2
XM_011538633.2 3000 UTR 3 XP_011536935.2
XM_011538634.2 3000 Intron XP_011536936.2
XM_011538635.2 3000 Intron XP_011536937.1
XM_011538636.2 3000 Intron XP_011536938.1
XM_017019774.1 3000 UTR 3 XP_016875263.1

View Full Product Details