Product Details

SNP ID
rs78937223
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:118156576 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGACTTTCCTTCTGAGGTCCAGA[C/T]GAGCTCCTGTAGTCTACCAGAGACA
Phenotype
MIM: 616711
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAOK3 PubMed Links

Gene Details

Gene
TAOK3
Gene Name
TAO kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016281.3 Intron NP_057365.3
XM_005253897.1 Intron XP_005253954.1
XM_005253898.1 Intron XP_005253955.1
XM_006719445.2 Intron XP_006719508.1
XM_011538437.1 Intron XP_011536739.1
XM_011538439.1 Intron XP_011536741.1
XM_017019408.1 Intron XP_016874897.1
XM_017019409.1 Intron XP_016874898.1
XM_017019410.1 Intron XP_016874899.1
XM_017019411.1 Intron XP_016874900.1
XM_017019412.1 Intron XP_016874901.1
XM_017019413.1 Intron XP_016874902.1

View Full Product Details