Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001160100.1 | 6073 | UTR 3 | NP_001153572.1 | ||
NM_006984.4 | 6073 | UTR 3 | NP_008915.1 | ||
NM_182848.3 | 6073 | UTR 3 | NP_878268.1 | ||
XM_011521134.2 | 6073 | Intron | XP_011519436.1 | ||
XM_017020843.1 | 6073 | Intron | XP_016876332.1 | ||
XM_017020844.1 | 6073 | Intron | XP_016876333.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_014934.4 | 6073 | UTR 3 | NP_055749.1 | ||
NM_198968.3 | 6073 | UTR 3 | NP_945319.1 | ||
XM_005254032.1 | 6073 | Intron | XP_005254089.1 | ||
XM_006719916.1 | 6073 | Intron | XP_006719979.1 | ||
XM_006719917.2 | 6073 | Intron | XP_006719980.1 | ||
XM_006719918.1 | 6073 | Intron | XP_006719981.1 | ||
XM_006719920.1 | 6073 | Intron | XP_006719983.1 | ||
XM_017020444.1 | 6073 | Intron | XP_016875933.1 | ||
XM_017020445.1 | 6073 | Intron | XP_016875934.1 | ||
XM_017020446.1 | 6073 | Intron | XP_016875935.1 | ||
XM_017020447.1 | 6073 | Intron | XP_016875936.1 |