Product Details

SNP ID
rs79533486
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40896013 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCATTGACTTCACCCCTTCCGAGC[A/G]CATTACCAGTCTTGTCGTCTCCAGC
Phenotype
MIM: 608551
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VPS18 PubMed Links

Gene Details

Gene
VPS18
Gene Name
VPS18, CORVET/HOPS core subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020857.2 526 Missense Mutation CAC,CGC H56R NP_065908.1
XM_011521843.2 526 Intron XP_011520145.1
XM_011521844.2 526 UTR 5 XP_011520146.1
XM_017022447.1 526 UTR 5 XP_016877936.1

View Full Product Details