Product Details
- SNP ID
-
rs79505053
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:42413789 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTCCCACAAAAGCAGGAGTGCAG[A/T]AGTAAGTAATGACTTTCTCTCAGAT
- Phenotype
-
MIM: 114240
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CAPN3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs28364558] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CAPN3
- Gene Name
- calpain 3
There are no transcripts associated with this gene.
- Gene
- ZNF106
- Gene Name
- zinc finger protein 106
View Full Product Details