Product Details

SNP ID
rs79592696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42744633 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGAAAATCAGGCCAAGAAGCCCA[A/T]ATGTTGAACTAAAACACAGGAGAAG
Phenotype
MIM: 607465 MIM: 611695
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CDAN1 PubMed Links

Gene Details

Gene
CDAN1
Gene Name
codanin 1
There are no transcripts associated with this gene.

Gene
TTBK2
Gene Name
tau tubulin kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173500.3 5218 UTR 3 NP_775771.3
XM_005254171.4 5218 UTR 3 XP_005254228.1
XM_005254173.4 5218 UTR 3 XP_005254230.1
XM_006720402.3 5218 UTR 3 XP_006720465.1
XM_006720403.3 5218 UTR 3 XP_006720466.1
XM_017021950.1 5218 UTR 3 XP_016877439.1

View Full Product Details