Product Details

SNP ID
rs79506770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:44294149 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCACATACCCTACACCCAGTTTCA[C/T]TGTGACATACCCCTTTCTTACTTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CASC4 PubMed Links

Gene Details

Gene
CASC4
Gene Name
cancer susceptibility candidate 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138423.3 Intron NP_612432.2
NM_177974.2 Intron NP_816929.1
XM_017021880.1 Intron XP_016877369.1

View Full Product Details