Product Details

SNP ID
rs75156596
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:45114846 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGACAGGTAAGACTGTACAAGAG[A/C]CTCCATGAATAGTCGAATTGAGGCT
Phenotype
MIM: 606759 MIM: 612771 MIM: 612772
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
DUOX2 PubMed Links

Gene Details

Gene
DUOX2
Gene Name
dual oxidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014080.4 Intron NP_054799.4
XM_005254421.2 Intron XP_005254478.1
Gene
DUOXA1
Gene Name
dual oxidase maturation factor 1
There are no transcripts associated with this gene.

Gene
DUOXA2
Gene Name
dual oxidase maturation factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207581.3 Intron NP_997464.2
XM_017022180.1 Intron XP_016877669.1

View Full Product Details