Product Details

SNP ID
rs78257203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:45116158 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGTGCAGAATGGTTCGTGGGTAC[A/G]GTGAACACCAACACATCCTACAAAG
Phenotype
MIM: 606759 MIM: 612771 MIM: 612772
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUOX2 PubMed Links

Gene Details

Gene
DUOX2
Gene Name
dual oxidase 2
There are no transcripts associated with this gene.

Gene
DUOXA1
Gene Name
dual oxidase maturation factor 1
There are no transcripts associated with this gene.

Gene
DUOXA2
Gene Name
dual oxidase maturation factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207581.3 525 Silent Mutation ACA,ACG T80T NP_997464.2
XM_017022180.1 525 Silent Mutation ACA,ACG T80T XP_016877669.1

View Full Product Details