Product Details

SNP ID
rs80228061
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:51723276 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAATCAGGATTGCATAAGGAGATA[A/C]ACATTATTTTTAGCCTAAATATATA
Phenotype
MIM: 611796
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LYSMD2 PubMed Links

Gene Details

Gene
LYSMD2
Gene Name
LysM domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143917.1 872 UTR 3 NP_001137389.1
NM_153374.2 872 UTR 3 NP_699205.1
XM_017022057.1 872 UTR 3 XP_016877546.1
Gene
SCG3
Gene Name
secretogranin III
There are no transcripts associated with this gene.

View Full Product Details