Product Details

SNP ID
rs75182225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:31327726 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGAGGAAGCACAAGTGCCACTAC[A/G]CGGGCTGCGAGAAAGTTTACGGGAA
Phenotype
MIM: 605328
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLF13 PubMed Links

Gene Details

Gene
KLF13
Gene Name
Kruppel like factor 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302461.1 872 Missense Mutation ACG,GCG T172A NP_001289390.1
NM_015995.3 872 Missense Mutation ACG,GCG T172A NP_057079.2

View Full Product Details