Product Details
- SNP ID
-
rs77376353
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:31484266 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGCGTCGCCCCGCGGCCCACCGCC[C/T]TTCTCCGCCGGCGACGCGCCCGCTG
- Phenotype
-
MIM: 612024
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OTUD7A
PubMed Links
Gene Details
- Gene
- OTUD7A
- Gene Name
- OTU deubiquitinase 7A
There are no transcripts associated with this gene.
View Full Product Details