Product Details

SNP ID
rs77256183
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:66349072 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTATATTCTTACCATTATTGCTAA[C/T]AAAATCTTCATGTAAAATAGGGAGA
Phenotype
MIM: 610716
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCARNA14 PubMed Links

Gene Details

Gene
SCARNA14
Gene Name
small Cajal body-specific RNA 14
There are no transcripts associated with this gene.

Gene
TIPIN
Gene Name
TIMELESS interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289986.1 732 Intron NP_001276915.1
NM_017858.2 732 Intron NP_060328.2
XM_005254521.2 732 Missense Mutation ATT,GTT I155V XP_005254578.1
XM_005254522.4 732 Missense Mutation ATT,GTT I155V XP_005254579.1
XM_006720594.3 732 Missense Mutation ATT,GTT I155V XP_006720657.1
XM_011521743.1 732 Missense Mutation ATT,GTT I155V XP_011520045.1
XM_017022390.1 732 Missense Mutation ATT,GTT I155V XP_016877879.1
XM_017022391.1 732 Intron XP_016877880.1

View Full Product Details