Product Details

SNP ID
rs79898187
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:66549131 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCATTATTAATAAAATAGTGGCC[A/G]CTTAGGTAATACCCACTTTTATCTT
Phenotype
MIM: 609984
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LCTL PubMed Links

Gene Details

Gene
LCTL
Gene Name
lactase like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278562.1 2934 Intron NP_001265491.1
NM_207338.3 2934 Intron NP_997221.2
XM_017021999.1 2934 Intron XP_016877488.1
Gene
ZWILCH
Gene Name
zwilch kinetochore protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287821.1 2934 UTR 3 NP_001274750.1
NM_001287822.1 2934 UTR 3 NP_001274751.1
NM_001287823.1 2934 UTR 3 NP_001274752.1
NM_017975.4 2934 UTR 3 NP_060445.3

View Full Product Details