Product Details

SNP ID
rs80315297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:67826175 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAAGAACTACAGCTATAATGAGA[C/T]CCACAACCGCCGCGTGGCCCTGGGC
Phenotype
MIM: 611273
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101929076 PubMed Links

Gene Details

Gene
LOC101929076
Gene Name
uncharacterized LOC101929076
There are no transcripts associated with this gene.

Gene
SKOR1
Gene Name
SKI family transcriptional corepressor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258024.1 469 Missense Mutation ACC,ATC T302I NP_001244953.1
XM_017022173.1 469 Missense Mutation ACC,ATC T116I XP_016877662.1

View Full Product Details