Product Details

SNP ID
rs80167401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:70049813 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATGAGCGGGTCTGTGGGGGAACC[A/G]GAGCCATAAGCCTCCAATAAATCTA
Phenotype
MIM: 600190
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TLE3 PubMed Links

Gene Details

Gene
TLE3
Gene Name
transducin like enhancer of split 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105192.2 3746 UTR 3 NP_001098662.1
NM_001282979.1 3746 UTR 3 NP_001269908.1
NM_001282980.1 3746 UTR 3 NP_001269909.1
NM_001282981.1 3746 UTR 3 NP_001269910.1
NM_001282982.1 3746 UTR 3 NP_001269911.1
NM_005078.3 3746 UTR 3 NP_005069.2
NM_020908.2 3746 UTR 3 NP_065959.1
XM_005254622.4 3746 UTR 3 XP_005254679.1
XM_005254623.3 3746 UTR 3 XP_005254680.1
XM_005254625.3 3746 UTR 3 XP_005254682.1
XM_005254628.3 3746 UTR 3 XP_005254685.1
XM_005254633.2 3746 UTR 3 XP_005254690.1
XM_006720665.3 3746 UTR 3 XP_006720728.1
XM_011521976.2 3746 UTR 3 XP_011520278.1
XM_011521977.2 3746 UTR 3 XP_011520279.1
XM_011521978.2 3746 UTR 3 XP_011520280.1
XM_011521979.2 3746 UTR 3 XP_011520281.1
XM_011521980.2 3746 UTR 3 XP_011520282.1
XM_011521981.1 3746 Intron XP_011520283.1
XM_011521982.2 3746 UTR 3 XP_011520284.1
XM_011521983.2 3746 UTR 3 XP_011520285.1
XM_017022532.1 3746 Intron XP_016878021.1

View Full Product Details