Product Details

SNP ID
rs78551559
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:70664767 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGTAGGTGAGTTGGGAAAGGCC[A/G]TTGAGTGCCTGTTTTAATCTTTCCA
Phenotype
MIM: 612516
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UACA PubMed Links

Gene Details

Gene
UACA
Gene Name
uveal autoantigen with coiled-coil domains and ankyrin repeats
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008224.1 3972 Silent Mutation AAC,AAT N1323N NP_001008225.1
NM_018003.2 3972 Silent Mutation AAC,AAT N1336N NP_060473.2
XM_005254529.4 3972 Silent Mutation AAC,AAT N1312N XP_005254586.1
XM_011521752.2 3972 Silent Mutation AAC,AAT N1356N XP_011520054.2
XM_011521753.2 3972 Silent Mutation AAC,AAT N1345N XP_011520055.2
XM_011521754.2 3972 Silent Mutation AAC,AAT N1084N XP_011520056.1
XM_017022394.1 3972 Silent Mutation AAC,AAT N1325N XP_016877883.1
XM_017022395.1 3972 Silent Mutation AAC,AAT N1319N XP_016877884.1

View Full Product Details