Product Details

SNP ID
rs76502381
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:71812401 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTCCTCTCCATACTCCTCTTCC[C/T]CCCCCTGCGGCCTGGACAGCATCCA
Phenotype
MIM: 604875 MIM: 604485
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYO9A PubMed Links

Gene Details

Gene
MYO9A
Gene Name
myosin IXA
There are no transcripts associated with this gene.

Gene
NR2E3
Gene Name
nuclear receptor subfamily 2 group E member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014249.3 833 Missense Mutation CCC,TCC P213S NP_055064.1
NM_016346.3 833 Missense Mutation CCC,TCC P213S NP_057430.1

View Full Product Details