Product Details

SNP ID
rs75302174
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:72755305 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTCCATTCACTTTCAACTTTTC[A/C]ATTCCAGAGGAGAAATATGATCAAC
Phenotype
MIM: 611861
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ADPGK PubMed Links

Gene Details

Gene
ADPGK
Gene Name
ADP dependent glucokinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031284.4 Intron NP_112574.3
XM_006720712.3 Intron XP_006720775.1
XM_006720713.3 Intron XP_006720776.1
XM_011522102.2 Intron XP_011520404.1
XM_011522103.2 Intron XP_011520405.1
XM_011522104.2 Intron XP_011520406.1
XM_017022664.1 Intron XP_016878153.1

View Full Product Details