Product Details

SNP ID
rs74614863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:73998572 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAGTCACACAGCTGAGGACAAG[A/G]AGCTTCTGGGGCCTTGCAACTCTAA
Phenotype
MIM: 102578 MIM: 608326
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PML PubMed Links

Gene Details

Gene
PML
Gene Name
promyelocytic leukemia
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002675.3 Intron NP_002666.1
NM_033238.2 Intron NP_150241.2
NM_033239.2 Intron NP_150242.1
NM_033240.2 Intron NP_150243.2
NM_033244.3 Intron NP_150247.2
NM_033246.2 Intron NP_150249.1
NM_033247.2 Intron NP_150250.2
NM_033249.2 Intron NP_150252.1
NM_033250.2 Intron NP_150253.2
Gene
STOML1
Gene Name
stomatin like 1
There are no transcripts associated with this gene.

View Full Product Details