Product Details

SNP ID
rs79222517
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75477092 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATAGGTGAAACAATCCACAGACA[G/T]GGCTTGTAAAAACTTCTGAACCAAA
Phenotype
MIM: 600768
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PTPN9 PubMed Links

Gene Details

Gene
PTPN9
Gene Name
protein tyrosine phosphatase, non-receptor type 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002833.3 Intron NP_002824.1

View Full Product Details