Product Details
- SNP ID
-
rs79305633
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:33859700 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTTGACATTACCTTTTTCTTCTT[C/T]GTCATTGTCATCTTGCTGGCCATCA
- Phenotype
-
MIM: 605265
MIM: 180903
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AVEN
PubMed Links
Gene Details
- Gene
- AVEN
- Gene Name
- apoptosis and caspase activation inhibitor
- Gene
- RYR3
- Gene Name
- ryanodine receptor 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001036.4 |
14528 |
Silent Mutation |
TTC,TTT |
F4756F |
NP_001027.3 |
NM_001243996.2 |
14528 |
Silent Mutation |
TTC,TTT |
F4751F |
NP_001230925.1 |
XM_011521880.2 |
14528 |
Silent Mutation |
TTC,TTT |
F4745F |
XP_011520182.1 |
XM_017022468.1 |
14528 |
Silent Mutation |
TTC,TTT |
F4752F |
XP_016877957.1 |
XM_017022469.1 |
14528 |
Silent Mutation |
TTC,TTT |
F4751F |
XP_016877958.1 |
XM_017022470.1 |
14528 |
Silent Mutation |
TTC,TTT |
F4751F |
XP_016877959.1 |
XM_017022471.1 |
14528 |
Silent Mutation |
TTC,TTT |
F4751F |
XP_016877960.1 |
XM_017022472.1 |
14528 |
Silent Mutation |
TTC,TTT |
F4750F |
XP_016877961.1 |
XM_017022473.1 |
14528 |
Silent Mutation |
TTC,TTT |
F4746F |
XP_016877962.1 |
XM_017022474.1 |
14528 |
Silent Mutation |
TTC,TTT |
F4745F |
XP_016877963.1 |
XM_017022475.1 |
14528 |
Silent Mutation |
TTC,TTT |
F4723F |
XP_016877964.1 |
XM_017022476.1 |
14528 |
Silent Mutation |
TTC,TTT |
F4716F |
XP_016877965.1 |
XM_017022477.1 |
14528 |
Intron |
|
|
XP_016877966.1 |
View Full Product Details