Product Details

SNP ID
rs74778029
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:33972831 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCAATACCCTGATTCAGAGTATC[A/G]GTCATTTGTGACTGACCCTCCTAGA
Phenotype
MIM: 605265 MIM: 118496
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AVEN PubMed Links

Gene Details

Gene
AVEN
Gene Name
apoptosis and caspase activation inhibitor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020371.2 Intron NP_065104.1
XM_011521818.2 Intron XP_011520120.1
XM_011521819.2 Intron XP_011520121.1
XM_011521820.1 Intron XP_011520122.1
Gene
CHRM5
Gene Name
cholinergic receptor muscarinic 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320917.1 Intron NP_001307846.1
NM_012125.3 Intron NP_036257.1

View Full Product Details