Product Details
- SNP ID
-
rs77761510
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:36801541 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAAAACAAATCTCTACCTCATACG[C/T]TCACTTAGACCCGTTGCTTCCCTCA
- Phenotype
-
MIM: 615626
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C15orf41
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs145479619] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C15orf41
- Gene Name
- chromosome 15 open reading frame 41
- Gene
- CSNK1A1P1
- Gene Name
- casein kinase 1 alpha 1 pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details