Product Details

SNP ID
rs78468046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:98437577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTTTCCTGAGGCCAGTATCCATA[A/C]CTCACTTATATCCCCTGTCCCCAGC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM169B PubMed Links

Gene Details

Gene
FAM169B
Gene Name
family with sequence similarity 169 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182562.2 2883 UTR 3 NP_872368.2

View Full Product Details