Product Details

SNP ID
rs77124139
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:99136940 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCCCATCAATACTGGAGGTGGT[A/G]CCCTGCACAAATGGCCCCAGGGTGG
Phenotype
MIM: 606087
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SYNM PubMed Links

Gene Details

Gene
SYNM
Gene Name
synemin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015286.5 2806 Intron NP_056101.5
NM_145728.2 2806 Intron NP_663780.2
XM_017022035.1 2806 Intron XP_016877524.1
Gene
TTC23
Gene Name
tetratricopeptide repeat domain 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288615.1 2806 UTR 3 NP_001275544.1
NM_001288616.1 2806 UTR 3 NP_001275545.1
XM_011521927.2 2806 UTR 3 XP_011520229.1
XM_011521929.1 2806 UTR 3 XP_011520231.1
XM_011521930.1 2806 UTR 3 XP_011520232.1
XM_011521931.1 2806 UTR 3 XP_011520233.1
XM_011521932.1 2806 UTR 3 XP_011520234.1
XM_011521933.1 2806 UTR 3 XP_011520235.1
XM_011521934.1 2806 UTR 3 XP_011520236.1
XM_011521935.1 2806 UTR 3 XP_011520237.1
XM_011521936.1 2806 UTR 3 XP_011520238.1
XM_011521937.1 2806 UTR 3 XP_011520239.1
XM_011521939.1 2806 UTR 3 XP_011520241.1
XM_011521940.2 2806 UTR 3 XP_011520242.1
XM_011521941.2 2806 UTR 3 XP_011520243.1
XM_011521943.2 2806 Intron XP_011520245.1
XM_011521944.2 2806 Intron XP_011520246.1
XM_011521945.2 2806 Intron XP_011520247.1
XM_017022509.1 2806 UTR 3 XP_016877998.1
XM_017022510.1 2806 UTR 3 XP_016877999.1
XM_017022511.1 2806 UTR 3 XP_016878000.1
XM_017022512.1 2806 UTR 3 XP_016878001.1
XM_017022513.1 2806 UTR 3 XP_016878002.1
XM_017022514.1 2806 UTR 3 XP_016878003.1
XM_017022515.1 2806 UTR 3 XP_016878004.1
XM_017022516.1 2806 UTR 3 XP_016878005.1
XM_017022517.1 2806 UTR 3 XP_016878006.1
XM_017022518.1 2806 UTR 3 XP_016878007.1

View Full Product Details