Product Details

SNP ID
rs77322249
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:101653982 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTTTTTTCTAAAGCGGAACAAA[A/G]CCTAATTATGAATAAGAAACATTTA
Phenotype
MIM: 610014
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TARSL2 PubMed Links

Gene Details

Gene
TARSL2
Gene Name
threonyl-tRNA synthetase like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152334.2 2514 UTR 3 NP_689547.2
XM_006720386.3 2514 UTR 3 XP_006720449.1
XM_017021912.1 2514 Intron XP_016877401.1
XM_017021913.1 2514 UTR 3 XP_016877402.1
Gene
TM2D3
Gene Name
TM2 domain containing 3
There are no transcripts associated with this gene.

View Full Product Details