Product Details

SNP ID
rs79102063
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:89582681 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCTAAGGTTGTTTGTCGTTTTAT[C/T]TTAGTTGTGGGAATCCCCAGACCAC
Phenotype
MIM: 613298
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TICRR PubMed Links

Gene Details

Gene
TICRR
Gene Name
TOPBP1 interacting checkpoint and replication regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308025.1 Intron NP_001294954.1
NM_152259.3 Intron NP_689472.3

View Full Product Details