Product Details

SNP ID
rs75454108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:89664652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATACACACGTGCCTGCAGGTGCA[C/T]AGCCCTGCATACACAAGAGATGGCA
Phenotype
MIM: 611254 MIM: 170290
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF7 PubMed Links

Gene Details

Gene
KIF7
Gene Name
kinesin family member 7
There are no transcripts associated with this gene.

Gene
PLIN1
Gene Name
perilipin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145311.1 2681 UTR 3 NP_001138783.1
NM_002666.4 2681 UTR 3 NP_002657.3
XM_005254934.4 2681 UTR 3 XP_005254991.1

View Full Product Details