Product Details

SNP ID
rs78188422
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:89735452 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGATATCAAAACTTTTAAACTCT[C/T]AAAGTAGGAGAATGTCTGTATATTT
Phenotype
MIM: 608689
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MESP1 PubMed Links

Gene Details

Gene
MESP1
Gene Name
mesoderm posterior bHLH transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018670.3 Intron NP_061140.1
Gene
WDR93
Gene Name
WD repeat domain 93
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284395.1 Intron NP_001271324.1
NM_001284396.1 Intron NP_001271325.1
NM_020212.1 Intron NP_064597.1
XM_006720614.1 Intron XP_006720677.1
XM_006720615.1 Intron XP_006720678.1
XM_006720618.2 Intron XP_006720681.1
XM_011521794.2 Intron XP_011520096.1
XM_011521795.1 Intron XP_011520097.1
XM_011521796.2 Intron XP_011520098.1
XM_011521798.2 Intron XP_011520100.1
XM_011521800.1 Intron XP_011520102.1
XM_011521802.2 Intron XP_011520104.1
XM_017022415.1 Intron XP_016877904.1
XM_017022416.1 Intron XP_016877905.1
XM_017022417.1 Intron XP_016877906.1

View Full Product Details