Product Details

SNP ID
rs76993218
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:20311280 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACCATAGGCCAGGCCACCAGGAA[A/C]CCTGAAATACAAGAAATATGACTGT
Phenotype
MIM: 602977
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GP2 PubMed Links

Gene Details

Gene
GP2
Gene Name
glycoprotein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007240.2 1491 Silent Mutation GGG,GGT G519G NP_001007241.2
NM_001007241.2 1491 Silent Mutation GGG,GGT G372G NP_001007242.2
NM_001007242.2 1491 Silent Mutation GGG,GGT G369G NP_001007243.2
NM_001502.3 1491 Silent Mutation GGG,GGT G516G NP_001493.2
XM_017023155.1 1491 Silent Mutation GGG,GGT G497G XP_016878644.1

View Full Product Details