Product Details

SNP ID
rs76483108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:20905156 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTATTCAGACAAAGCTTAGCATAG[A/G]TAAAAGAGACCTCGTTGTTCTTCCT
Phenotype
MIM: 616167 MIM: 614709
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCUN1D3 PubMed Links

Gene Details

Gene
DCUN1D3
Gene Name
defective in cullin neddylation 1 domain containing 3
There are no transcripts associated with this gene.

Gene
LYRM1
Gene Name
LYR motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128301.2 Intron NP_001121773.1
NM_001128302.2 Intron NP_001121774.1
NM_001302835.1 Intron NP_001289764.1
NM_001302836.1 Intron NP_001289765.1
NM_020424.4 Intron NP_065157.1
XM_005255444.1 Intron XP_005255501.1
XM_011545900.2 Intron XP_011544202.1
XM_011545901.2 Intron XP_011544203.1
XM_011545902.1 Intron XP_011544204.1
XM_011545908.2 Intron XP_011544210.1
XM_017023475.1 Intron XP_016878964.1
XM_017023476.1 Intron XP_016878965.1
XM_017023477.1 Intron XP_016878966.1
XM_017023478.1 Intron XP_016878967.1
XM_017023479.1 Intron XP_016878968.1
XM_017023480.1 Intron XP_016878969.1
XM_017023481.1 Intron XP_016878970.1
XM_017023482.1 Intron XP_016878971.1

View Full Product Details