Product Details
- SNP ID
-
rs74640315
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:21317025 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTTTCTACATCACAGCTTCCTTAA[C/T]TTCTCCTTCTCCTTTGGAATTGGCT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CRYM-AS1
PubMed Links
Gene Details
- Gene
- CRYM-AS1
- Gene Name
- CRYM antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details