Product Details

SNP ID
rs76820730
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2362437 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTACTTCCTAGAAAGCAGCTAC[A/G]TGAGGTTTTCTCCCAATAGGGCCTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCA17P PubMed Links

Gene Details

Gene
ABCA17P
Gene Name
ATP binding cassette subfamily A member 17, pseudogene
There are no transcripts associated with this gene.

View Full Product Details