Product Details
- SNP ID
-
rs77689392
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:30413091 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTATTTTATTAATTAGAGAAGTAAT[A/G]GCTAAGGAAGAAACTGGGATGTTTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF771
PubMed Links
Gene Details
- Gene
- ZNF771
- Gene Name
- zinc finger protein 771
View Full Product Details