Product Details

SNP ID
rs77284397
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31061385 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTGCACTCTCGGCACACAAACT[G/T]GGGGGGCTTCTCGTCAGCCTCCTCA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF668 PubMed Links

Gene Details

Gene
ZNF668
Gene Name
zinc finger protein 668
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172668.1 1854 Missense Mutation AAG,CAG K515Q NP_001166139.1
NM_001172669.1 1854 Missense Mutation AAG,CAG K538Q NP_001166140.1
NM_001172670.1 1854 Missense Mutation AAG,CAG K515Q NP_001166141.1
NM_024706.4 1854 Missense Mutation AAG,CAG K515Q NP_078982.3

View Full Product Details