Product Details

SNP ID
rs74947798
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3137631 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCGGGAAGCGGTGAGGAGGCTGT[C/T]GCCTTGGTGGAGGACCTACAGAAGC
Phenotype
MIM: 608387
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CASP16P PubMed Links

Gene Details

Gene
CASP16P
Gene Name
caspase 16, pseudogene
There are no transcripts associated with this gene.

Gene
ZNF213
Gene Name
zinc finger protein 213
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134655.1 766 Silent Mutation GTC,GTT V117V NP_001128127.1
NM_004220.2 766 Silent Mutation GTC,GTT V117V NP_004211.1
XM_011522652.2 766 Silent Mutation GTC,GTT V117V XP_011520954.1
Gene
ZNF213-AS1
Gene Name
ZNF213 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details