Product Details

SNP ID
rs77375403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3310825 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTTCCCTTAATGCTTGTTCCTTG[A/G]CTCTGCTGACAAGTTGGGAGCCAGG
Phenotype
MIM: 612969 MIM: 604191 MIM: 601473
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TIGD7 PubMed Links

Gene Details

Gene
TIGD7
Gene Name
tigger transposable element derived 7
There are no transcripts associated with this gene.

Gene
ZNF263
Gene Name
zinc finger protein 263
There are no transcripts associated with this gene.

Gene
ZNF75A
Gene Name
zinc finger protein 75a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302109.1 215 Intron NP_001289038.1
NM_001324041.1 215 Intron NP_001310970.1
NM_001324085.1 215 Intron NP_001311014.1
NM_153028.3 215 Intron NP_694573.1
XM_011522646.2 215 Intron XP_011520948.1
XM_011522647.2 215 UTR 5 XP_011520949.1
XM_017023642.1 215 Intron XP_016879131.1
XM_017023643.1 215 Intron XP_016879132.1
XM_017023644.1 215 Intron XP_016879133.1
XM_017023645.1 215 Intron XP_016879134.1

View Full Product Details