Product Details
- SNP ID
-
rs76808454
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:3371892 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAAAGTAAGAGACAGCTGAACCCT[C/T]GTGTGGCCATTCATACAAGTCCCTA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MTRNR2L4
PubMed Links
Gene Details
- Gene
- MTRNR2L4
- Gene Name
- MT-RNR2-like 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001190476.1 |
392 |
Missense Mutation |
CAA,CGA |
Q4R |
NP_001177405.1 |
View Full Product Details