Product Details

SNP ID
rs78010013
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3542637 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCATTTTATTCCATCAGGGAGGA[A/C]CCAGCAGAGAACTCTGCTGCTCCAG
Phenotype
MIM: 616787 MIM: 615648
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CLUAP1 PubMed Links

Gene Details

Gene
CLUAP1
Gene Name
clusterin associated protein 1
There are no transcripts associated with this gene.

Gene
LOC101929732
Gene Name
uncharacterized LOC101929732
There are no transcripts associated with this gene.

Gene
NLRC3
Gene Name
NLR family CARD domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178844.3 Intron NP_849172.2
XM_017023027.1 Intron XP_016878516.1
XM_017023028.1 Intron XP_016878517.1
XM_017023029.1 Intron XP_016878518.1
XM_017023030.1 Intron XP_016878519.1
XM_017023031.1 Intron XP_016878520.1
XM_017023032.1 Intron XP_016878521.1
XM_017023033.1 Intron XP_016878522.1
XM_017023034.1 Intron XP_016878523.1
XM_017023035.1 Intron XP_016878524.1
XM_017023036.1 Intron XP_016878525.1
XM_017023037.1 Intron XP_016878526.1
XM_017023038.1 Intron XP_016878527.1
XM_017023039.1 Intron XP_016878528.1

View Full Product Details