Product Details

SNP ID
rs79642041
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4260216 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGTGCTGGCACGATCACCGTGGG[A/G]TGGTGGGTGGGGGCCGGAGGGGGCA
Phenotype
MIM: 600743
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LINC01569 PubMed Links
Additional Information
For this assay, SNP(s) [rs528385650] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
LINC01569
Gene Name
long intergenic non-protein coding RNA 1569
There are no transcripts associated with this gene.

Gene
LOC105371063
Gene Name
putative uncharacterized protein encoded by LINC00596
There are no transcripts associated with this gene.

Gene
TFAP4
Gene Name
transcription factor AP-4 (activating enhancer binding protein 4)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003223.2 1086 Silent Mutation CAC,CAT H232H NP_003214.1
XM_011522633.2 1086 Silent Mutation CAC,CAT H219H XP_011520935.1
XM_011522635.2 1086 Silent Mutation CAC,CAT H172H XP_011520937.1

View Full Product Details