Product Details

SNP ID
rs77302158
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4746562 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCAGCCTCTACTTTGCAGAGTTG[C/T]CTGTTGACCGCACAGAGCCTCTGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C16orf71 PubMed Links

Gene Details

Gene
C16orf71
Gene Name
chromosome 16 open reading frame 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139170.2 Intron NP_631909.2
XM_005255144.3 Intron XP_005255201.1
XM_017022975.1 Intron XP_016878464.1
XM_017022976.1 Intron XP_016878465.1
Gene
ZNF500
Gene Name
zinc finger protein 500
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303450.1 Intron NP_001290379.1
NM_021646.3 Intron NP_067678.1
XM_005255243.3 Intron XP_005255300.1
XM_011522453.2 Intron XP_011520755.1
XM_011522454.2 Intron XP_011520756.1
XM_017023121.1 Intron XP_016878610.1

View Full Product Details